Microbiologische media, kleuringen en toebehoren
BD Difco™ Dehydrated Culture Media: Leptospira Medium Base EMJH
Use with Leptospira Enrichment EMJH to cultivate and maintain Leptospira
BD Difco™ Dehydrated Culture Media: ISP Medium 2
International Streptomyces Project Yeast Malt Extract Agar
Thermo Scientific™ Oxoid™ AnaeroGen™-sachet, 3.5 L
Thermo Scientific™ Oxoid AnaeroGen 3.5L Sachet is for use with Thermo Scientific™ Oxoid Anaerobic 3.5L jars, Part No. HP0011A, HP0031A.
Thermo Scientific™ Oxoid™ technische agar
The Oxoid Agar Technical (Agar No. 3) is a general-purpose bacteriological-grade agar.
BD Difco™ Columbia-agar met 5% schapenbloed
Recommended when processing clinical specimens for unusual organisms. BD™ BBL™ Columbia Agar with 5% Sheep Blood is a highly nutritious general purpose medium for the isolation and cultivation of nonfastidious and fastidious microorganisms from clinical specimens.
Thermo Scientific™ Oxoid™ MRS-bouillon
Isolate and enumerate lactic acid bacteria from food with this non-selective medium.
Thermo Scientific™ Brilliance™ Salmonella Agar Base
Isolate and presumptively identify Salmonella species from food samples with this chromogenic medium.
Thermo Scientific™ Oxoid™ Resazurin anaërobe indicator
Oxoid Resazurin is an indicator strip for use with anaerobic jar systems.
Thermo Scientific™ Oxoid™ MRS-agar
Isolate and enumerate lactic acid bacteria from food samples with this medium.
BD Difco™ gedehydrateerd kweekmedium: M 17 Broth
Selective isolation and enumeration of streptococci. BD Difco™ Dehydrated Culture Media: M 17 Broth is part of a large selection of dehydrated media.
BD Difco™ Dehydrated Culture Media: Malt Extract Agar
For isolation, detection and enumeration of yeasts and molds
BD Difco™ gedehydrateerd kweekmedium: Gist-/schimmelbouillon
For cultivation of yeasts, molds and other aciduric microorganisms. BD Difco™ Dehydrated Culture Media: Yeast Mold Broth is part of a large selection of dehydrated media.
BD Difco™ Leptospira Enrichment EMJH
Used with Leptospira Medium Base EMJH in cultivating Leptospira. BD™ Difco™ Leptospira Enrichment EMJH is a lyophilized media additive. The solution contains albumin, polysorbate 80 and additional growth factors for Leptospira.
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Rosolic Acid
Use with mFC Broth Base and mFC Agar for detection and enumeration of fecal coliforms
Thermo Scientific™ Oxoid™ CampyGen™-sachet, 2.5 L
Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with Oxoid CampyGen 2.5L Sachet.
Fisher BioReagents™ Additieven voor microbiologische media: Casaminozuren
Additive for preparing bacterial nutrient media
BD Difco™ Dehydrated Culture Media: m Enterococcus Agar
For isolating and enumerating enterococci in water and other materials by membrane filtration or pour plate technique
BD GasPak™ verbruiksartikelen
Accessoires en toebehoren voor gebruik met BD™ Diagnostic Systems GasPak™ gasproducerende producten
BD BBL™ MGIT™ indicatorbuis voor mycobacteriegroei: reageermiddelen
Offers superior detection time in a user-friendly, fluorescent test format
Thermo Scientific™ Oxoid™ tabletten met Ringer-oplossing
Oxoid Ringers Solution Tablets is used for the preparation of isotonic diluents for bacterial cells and bacteriological specimens.
Thermo Scientific™ Oxoid™-tryptoonsoja-agar
Grow a wide variety of microorganisms with this general purpose medium specified in international pharmacopoeia.
Fisher BioReagents™ Agar, Powder / Flakes, Fisher BioReagents™
Used as a solidifying agent for culture media
Thermo Scientific™ Oxoid™ tabletten voor fosfaatgebufferde zoutoplossing
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.
Thermo Scientific™ Oxoid™ aardappeldextrose-agar
Isolate and enumerate yeasts and molds from dairy and food products with this medium while conforming to EP/USP/JP Microbial Limit Test standards.
Thermo Scientific™ Oxoid™ AnaeroGen™-sachet, 2.5 L
Thermo Scientific™ Oxoid AnaeroGen 2.5L Sachets are anaerobic gas generating sachets for use with Thermo Scientific™ Oxoid 2.5L jar, Part No. AG0025A (not included).
Thermo Scientific™ Oxoid™ Wilkins-Chalgren anaerobe bouillon
Isolate, cultivate and /or carry out susceptibility testing of a wide variety of anaerobic organisms from clinical samples with this medium.
BD Difco™ gedehydrateerd kweekmedium: Korrelvormige agar
Gebruikt als stollingsmiddel voor cultuurmedia. Gedehydrateerde BD Difco™-cultuurmedia: Korrelvormige agar is zorgvuldig gecontroleerd op cultuurrespons, oplosbaarheid en gelatineringstemperatuur.
BD Difco™ Dehydrated Culture Media: Mitis Salivarius Agar
Use with Chapman Tellurite Solution 1% for selective isolation of S. mitis, S. salivarius and enterococci
BD Difco™ gedehydrateerd kweekmedium: Gist op stikstofbasis zonder aminozuren
Used for classifying yeasts based on amino acid and carbohydrate requirements. BD™ Difco™ Yeast Nitrogen Base without Amino Acids is a dehydrated culture medium used in many molecular genetics applications. Prepared according to the formulas of Wickerham.
BD BBL™ gedehydrateerd kweekmedium: Brucella-bouillon
In gedehydrateerde vorm geleverd voor het gemak van de laborant en de bioverwerkingsspecialist. BBL™ gedehydrateerde BD Difco™-cultuurmedia: Brucella Broth behoort tot een uitgebreide selectie van gedehydrateerde media.