TLC-producten
DWK Life Sciences Kimble™ Kontes™ Chromatography Cylindrical TLC Developing Tanks
DWK Life Sciences Kimble™ Kontes™ Chromatography Cylindrical TLC Developing Tanks
Cylindrical glass tanks for the development of 1 x 3 in. to 10 x 20cm plates.
DWK Life Sciences Kontes™ rechthoekige TLC ontwikkeltanks met deksel
DWK Life Sciences Kontes™ rechthoekige TLC ontwikkeltanks met deksel
Molded glass tanks for development of any size TLC plate up to 20 × 20cm
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers Head Only
DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers Head Only
TLC Sprayer head for use with 125 mL Flask with Knots Sprayer System
DWK Life Sciences Kimble™ Chromatography TLC Plate Streaker
DWK Life Sciences Kimble™ Chromatography TLC Plate Streaker
A simple device for uniformly streaking TLC plates.
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Kimble Chase™ Drummond Microcaps Borosilicate Glass Capillary Tubes
Kimble Chase Drummond Microcaps Borosilicate Glass Capillary Tubes, Capacity: 0.5 uL, Length: 32 mm, Quantity: 100/Pk.
DWK Life Sciences Kimble™ Kontes™ Replacement Parts for TLC Equilibration Apparatus
DWK Life Sciences Kimble™ Kontes™ Replacement Parts for TLC Equilibration Apparatus
For use with Kontes™ rectangular developing tank (K416190-4900)
DWK Life Sciences Kontes™ reagenssproeiers met verbindingshuls met schroefdraad
DWK Life Sciences Kontes™ reagenssproeiers met verbindingshuls met schroefdraad
Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids
DWK Life Sciences Kimble™ Kontes™ TLC Cylindrical Developing Tanks
DWK Life Sciences Kimble™ Kontes™ TLC Cylindrical Developing Tanks
Cylindrical glass tanks for the development of 5 × 10cm (2 × 3.9 in. ) and 5 × 20cm (2 × 7.9 in. ) plates
MP Biomedicals™ EcoChrome™ Silica TLC Plate
MP Biomedicals™ EcoChrome™ Silica TLC Plate
Used as a defoamer component. In hydrated form, it is used in toothpaste as a hard abrasive to remove tooth plaque. SiO
Macherey-Nagel™ Standard SILGUR Silica Layers on Glass Plates
Macherey-Nagel™ Standard SILGUR Silica Layers on Glass Plates
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SILGUR silica layers on glass plates come in a variety of plate sizes and layer thicknesses.  Select a fluorescent indicator and choose from a variety of pack sizes.
Macherey-Nagel™ Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates
Macherey-Nagel™ Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates
Separate mixtures with sharp separations, short developing times and short migration distances. Nano-SIL Unmodified HPTLC Silica Layers on Glass Plates have an increased detection sensitifvity compared to SIL G plates.
Macherey-Nagel™ Glazen plaat, adamant UV254
Macherey-Nagel™ Glazen plaat, adamant UV254
Increase separation efficiency and maximize resolution during TLC (thin-layer chromatography). Adamant™ Unmodified Standard Silica Layers for TLC, UV254, are available on glass plates with an optimized binder system and UV indicator.
Macherey-Nagel™ Glasplaten, silica
Macherey-Nagel™ Glasplaten, silica
Gescheiden niet-vluchtige mengsels met ongemodificeerde standaard silicalagen voor TLC (dunnelaagchromatografie). Standaard SIL G silicalagen op glazen platen zijn verkrijgbaar in verschillende plaatafmetingen en laagdikten. Selecteer een fluorescentie-indicator en kies uit de verschillende verpakkingsformaten.
Macherey-Nagel™ Alugram-aluminium plaat SIL G
Macherey-Nagel™ Alugram-aluminium plaat SIL G
Gescheiden niet-vluchtige mengsels met ongemodificeerde standaard silicalagen voor TLC (dunnelaagchromatografie). Standaard SIL G silicalagen op Alugram™-aluminiumplaten zijn verkrijgbaar in meerdere afmetingen.
Macherey-Nagel™ Chiralplate™ for TLC Enantiomer Separation
Macherey-Nagel™ Chiralplate™ for TLC Enantiomer Separation
Resolve a wide variety of enantiomers. Chiralplate™ for TLC Enantiomer Separation consists of a special layer available on glass plates with fluorescent indicator.
Macherey-Nagel™ Polygram SIL G Polyester Sheets
Macherey-Nagel™ Polygram SIL G Polyester Sheets
Macherey-Nagel Polygram SIL G Polyester Sheets, Dimensions: 40 x 20 cm, Pore Size: 60 A, Pore Volume: 0.75 mL/g, Quantity: 25 Pack, Thickness: 0.2 mm
Macherey-Nagel™ SIL G-25 Tenside Special Silica Layers for Separation of Surfactant
Macherey-Nagel™ SIL G-25 Tenside Special Silica Layers for Separation of Surfactant
 Separate surfactants by TLC (Thin-Layer Chromatography).  SIL G-25 Tenside Special Silica Layers for Separation of Surfactant come avallable as glass plates.
  spinner