Enzymen en remmers
Thermo Scientific™ 10X TAE-buffer (Tris-boraat-EDTA)
Perform electrophoresis of nucleic acids in agarose and polyacrylamide gels using Thermo Scientific™ 10X TBE Buffer (Tris-borate-EDTA).
Thermo Scientific™ FastDigest BsiWI (Pfl23II)
Cut at C^GTACG sites with Thermo Scientific™ FastDigest™ BsiWI (Pfl23II), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: PspLI).
Thermo Scientific™ Cfr9I (XmaI)
The Cfr9I (XmaI) restriction enzyme recognizes C^CCGGG sites and cuts best at 37°C in its own unique buffer (Isoschizomers: TspMI, XmaCI, XmaI).
Thermo Scientific™ FastDigest™ EcoRI
The FastDigest EcoRI restriction enzyme recognizes G^AATTC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest BbsI (BpiI)
Cut at GAAGAC(2/6)^ sites with Thermo Scientific™ FastDigest™ BbsI (BpiI), which performs best using universal FastDigest Buffer (Isoschizomers: BpuAI, BstV2I).
Thermo Scientific™ FastDigest NotI
The FastDigest NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: CciNI).
Thermo Scientific™ FastDigest Bsp1407I
Cut at T^GTACA sites with Bsp1407I restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest™ Buffer (Isoschizomers: BsrGI, BstAUI).
Invitrogen™ Anza™ 29 KflI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest PvuII
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest SanDI (KflI)
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest™ BamHI
Cut at G^GATCC sites with Thermo Scientific™ FastDigest™ BamHI restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Invitrogen™ Anza™ 3 BcuI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 4 BpiI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest MunI
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest SexAI (CsiI)
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest AflII (BspTI)
Cut at C^TTAAG sites with Thermo Scientific™ FastDigest™ AflII (BspTI), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.
Thermo Scientific™ FastDigest Eco32I
Cut at GAT^ATC sites with Thermo Scientific™ FastDigest™ EcoRV (Eco32I), which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: EcoRV).
Thermo Scientific™ FastDigest PsiI (AanI)
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ FastDigest Bsu15I
Cut at AT^CGAT sites with ClaI (Bsu15I) restriction enzyme, which performs best at 37°C in 5-15 minutes using universal FastDigest™ Buffer.
Invitrogen™ Anza™ 8 XhoI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Invitrogen™ Anza™ 21 SgsI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ AarI (2 U/µL)
Cut at CACCTGC(4/8)^ sites with AarI restriction enzyme, which performs best at 37°C in its own unique (+oligo) buffer.
Thermo Scientific™ FastDigest SapI (LguI)
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ Anza™ 27 PvuI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ FastDigest NdeI
The FastDigest NdeI restriction enzyme recognizes CA^TATG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: FauNDI).
Thermo Scientific™ FastDigest MluI
Perform single-, double- or multiple DNA digestion within 5-15 minutes withthis advanced line of restriction enzymes, 100% active in one universal buffer.
Invitrogen™ Anza™ 10 DpnI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Thermo Scientific™ KpnI
The KpnI restriction enzyme recognizes GGTAC^C sites and cuts best at 37°C in its own unique buffer (Isoschizomers: Asp718I).
Thermo Scientific™ FastDigest BoxI
Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.
Thermo Scientific™ Eco52I (EagI) (10 U/µL)
The Eco52I (EagI) restriction enzyme recognizes C^GGCCG sites and cuts best at 37°C in its own unique buffer (Isoschizomers: BseX3I, BstZI, EagI, EclXI).