Roerders en roerstaven
IKA™ IKA™ RCT Basic IKAMAG™ veiligheidsbediening magneetroerder met warmteplaat
Stirring Range: 50 to 1500 rpm; Size: 5.3 in. dia.
Heidolph instruments MR Series Magnetic Hot Plate Stirrer with Pt1000 Temperature Sensor, 20°C to 300°C, Silumin
Stirring Range: 100 to 1400 rpm; Shape: Round; Size: 145 mm dia.; Max. Sample Load: 25 kg
Fisherbrand™ Warmteplaatroerder model ARE, aluminium
Stirring Range: 1500 rpm; Size: 135 mm dia.
Fisherbrand™ PTFE-roerstaven met vleugel
Voor stabiel roeren. De PTFE-roerstaven met vleugel zijn effectief in kleine, tapse reageer-, centrifugeer- en monsterbuizen met ronde bodem.
IKA™ RH Digital Magnetic Stirrer
An innovatively designed digital magnetic heated stirrer with bushing according to DIN 12878 for connecting an electronic contact thermometer, such as the ETS-D5.
Fisherbrand™ Octagon Spinbar™ Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape for stirring small volumes in vessels such as vials, tubes and gradient makers.
BRAND™ Cilindervormige, magnetische PTFE-roerstaven
Vervaardigd van PTFE. BRAND™ PTFE magnetische roerstaven met cilindervorm zijn geschikt voor universele toepassingen. Afgerond, glad oppervlak zorgt voor effectief roeren.
IKA™ IKA RET Basic IKAMAG™ magneetroerder
Set exact temperature and speed setting via digital display, even when switched off. IKA™ RET Basic Magnetic Stirrer includes a wide speed range from 0/50 to 1700rpm and integrated temperature control. Hot top indicator warns to prevent burns.
IKA™ C-MAG MS 7 Magnetic Stirrer
Use this magnetic stirrer without heating for a variety of applications. IKA™ C-MAG MS 7 Magnetic Stirrer features a ceramic set-up plate which offers excellent chemical resistance.
IKA™ RCT Digital Magnetic Stirring Hotplate
RCT digital stirrer has a round top made of aluminum alloy, stirs volumes of up to 20 L (H2O), and runs at speeds ranging from 50 to1500 rpm.
Velp Scientifica™ Aluminium roerder met verwarmingsplaat model AM4
Multiple-position hot plate stirrer with four separately controlled stirring plates.
Thermo Scientific™ Cimarec i Poly 15 and Multipoint Stirrers
Conduct stirring applications with these stirrers, which are 100% maintenance- and wear-free, in a fully synchronized multipoint platform.
Thermo Scientific™ RT Basic Series Magnetic Stirrers
Experience the reliability of our plug-and-play magnetic stirrers, ideal for routine stirring applications.
Thermo Scientific™ Cimarec™ Mono & Maxi Direct Stirrers
Ensure reliability, safety and performance with these mono and maxi direct stirrers, which are 100% maintenance- and wear-free.
Fisherbrand™ Isotemp™ Hot Plate Stirrer, Ambient to 300°C, Aluminum
Stirring Range: 50 to 1500 rpm; Shape: Square; Size: 7.25 x 7.25 in.; Plug Type: EU-Style
Fisherbrand™ Cilindrische roerstaven van PTFE, set in doos
Provides general purpose stirring for a wide variety of applications. Fisherbrand™ Stir Bar Box Sets, Cylindrical are isostatically molded in pure, FDA-approved PTFE plastic.
Fisherbrand™ Cilindrische roerstaven van PTFE
Alnico V magnet core encapsulated by PTFE or glass . Fisherbrand™ Cylindrical PTFE or Glass Stir Bars are autoclavable.
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Thermo Scientific™ RT Touch Series Magnetic Stirrers
Stay in control with our low-profile, digital display stirrers, ideal for routine stirring applications where visible stir speed is required.
Heidolph instruments MR Hei-Tec Magnetic Stirrer with Heating
Includes leading safety standards and features for superior ease of use and reduced cost of ownership. Heidolph™ MR Hei-Tec Magnetic Stirrer with Heating includes a variety of features for easy monitoring and control.
IKA™ Keramische magneetroerder met warmteplaat
Stirring Range: 100 to 1500 rpm; Shape: Square; Size: 260 x 260 mm
IKA™ RT 15 Magnetic Hot Plate Stirrer, Ambient to 120°C, Aluminum Alloy
Stirring Range: 0 to 1000 rpm; Shape: Rectangle; Size: 10.6 x 19.5 in.; Plug Type: EU-Style
Fisherbrand™ Isotemp™ Hot Plate Stirrer, Ambient to 400°C, Ceramic
Stirring Range: 50 to 1500 rpm; Shape: Square; Size: 10.25 x 10.25 in.; Plug Type: EU-Style
Thermo Scientific™ Cimarec™ i Telesystem Multipoint Stirrers
Safely conduct applications with aggressive media in high-humidity environments with these multipoint stirrers.
Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars
Use these suspended magnetic stir bars for constant low-speed stirring action in tissue culture applications.
Thermo Scientific™ RT2 Advanced Hotplate Stirrer
Experience precision with our low-profile, digital display hotplate stirrer, equipped with PT100 sensor for precise temperature measurements for sensitive experimental procedures.
Fisherbrand™ PTFE Stir Bar with Removable Ring
Provides stable stirring action. Fisherbrand™ PTFE Stir Bar with Removable Ring are autoclavable and include a pivot ring.
Fisherbrand™ Isotemp™ Stirrer
Experience precise stirring control and exceptional safety performance for your routine protocols. Available in three sizes, which provide flexibility from microscale chemistries to production operations.
Fisherbrand™ Stir Bar Box Set, Cross-Shaped
Provides stable, general purpose stirring. Fisherbrand™ Stir Bar Box Sets, Cross-Shaped are isostatically molded in pure, FDA-approved PTFE plastic.