Maatcilinders
BRAND™ Short Form Polypropylene Graduated Cylinder
Polypropylene, high clarity.
Thermo Scientific™ Nalgene™ PMP hydrometerpot
Protect against breakage. This PMP jar is lighter than glass and offers superior chemical resistance and a broad base, making it a stable addition to the lab or field.
Cole Parmer™ Economical Polypropylene Graduated Cylinder
Polypropylene (PP) cylinders offer very good chemical resistance.
BRAND™ Tall Form Graduated Cylinder
Tall Form Graduated Cylinder is crystal clear and includes and embossed scale
BRAND™ Grote maatcilinders van polypropyleen, klasse B
Vervaardigd van zeer helder polypropyleen. Grote maatcilinders van polypropyleen, klasse B, met blauwe schaalopdruk, bieden eenvoudige volume-inschatting. Blootstelling aan temperaturen tot 80 °C veroorzaakt geen permanent overschreden foutlimieten. Reinigen tot 60 °C wordt aanbevolen voor behoud van markeringen en inscripties.
BRAND™ Low Form Graduated Cylinder
Low Form Graduated Cylinder designed for high clarity and visibility.
BRAND™ Class B PMP Tall Form Measuring Cylinders
Autoclaving up to 121°C will not cause permanently exceeded error limits. Cleaning up to max. 60°C is recommended to preserve marks and inscriptions.
Duran™ Ungraduated Measuring Cylinders
These measuring cylinders are for holding and measuring varying amounts of liquids in the laboratory.
Kartell™ Grote maatcilinders van doorzichtig polypropyleen met blauwe schaalverdeling, klasse B
Vervaardigd van zeer transparant polypropyleen voor duurzaamheid, helderheid en goede chemische bestendigheid. Autoclaveerbare Kartell™ hoge polypropyleen maatcilinders met blauwe schaalverdeling bieden een voordelige en veilige oplossing voor het meten van het volume van vloeistoffen in een laboratorium.
Bohlender™ BOLA™ Measuring Cylinders
Translucent measuring cylinders with hexagonal base to prevent rolling.
Kartell™ Tall Form Graduated Measuring Cylinders
Accurately measures the volume of a liquid. Kartell™ Tall Form Graduated Measuring Cylinders use horizontal marked lines to assist in measuring out various volumes of a liquid.
Hirschmann™ Class B Graduated Cylinders with Blue Graduations
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann™ Class B Measuring Cylinders, raised scale are manufactured from PP.