Celcultuurmedia
Gibco™ Opti-MEM™ I Reduced Serum Medium
Opti-MEM™ I Reduced-Serum Medium is an improved Minimal Essential Medium (MEM) that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in growth rate or morphology.
Gibco™ DMEM, no glucose
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, No Glucose is used in cell culture processing applications.
Gibco™ Cell Dissociation Buffer, enzyme-free, PBS
Animal origin-free
Gibco™ DMEM, high glucose, NEAA, no glutamine
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, NEAA, No Glutamine, Sodium Bicarbonate Buffer is used in human ex vivo tissue and cell culture processing applications.
Gibco™ Recovery™ Cell Culture Freezing Medium
Complete, ready-to-use freezing medium for cryopreservation of a wide variety of mammalian cells, including CHO-S, CHO-K1, HEK 293, Jurkat, and NIH 3T3
Gibco™ Opti-MEM™ I Reduced Serum Medium, no phenol red
Improved Minimal Essential Medium that allows for a reduction of Fetal Bovine Serum supplementation by at least 50% with no change in cell growth rate or morphology
Gibco™ RPMI 1640 Medium, HEPES
Originally developed to culture human leukemic cells in suspension and as monolayer
Gibco™ DMEM/F-12
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM/F-12 is used in cell culture processing applications.
Gibco™ Neurobasal™ Medium
Used in the growth of neuronal cells from hippocampus, cortex and otregions of the brain. Gibco™ Neurobasal™ Medium is a basal medium that meets the special cell culture requirements of pre-natal and embryonic neuronal cells when used with Gibco™ B-27™ Supplement.
Gibco™ Advanced RPMI 1640 Medium
Advanced RPMI (Roswell Park Memorial Institute) 1640 is widely used basal medium that allows culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation
Gibco™ DMEM, high glucose, GlutaMAX™ Supplement
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, GlutaMAX™ Supplement is used in human ex vivo tissue and cell culture processing applications.
Gibco™ Dispase II, powder
Amino-endo peptidase that hydrolyzes N-terminal peptide bonds of non-polar amino acid residues
Gibco™ IMDM
Iscove's Modified Dulbecco's Medium is well suited for rapidly proliferating, high-density cell cultures, including Jurkat, COS-7, and macrophage cells
Gibco™ DMEM/F-12, GlutaMAX™ supplement
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEMF/12, GlutaMAX™ Supplement is used in cell culture processing applications.
Gibco™ DMEM, high glucose, no glutamine, no phenol red
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, No Glutamine, No Phenol Red is used in cell culture processing applications.
Gibco™ MEM
Minimum Essential Medium is one of the most commonly used of all cell culture media
Gibco™ DMEM, high glucose, HEPES, no phenol red
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, HEPES, No Phenol Red is used in cell culture processing applications.
Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ Essential 8™ Medium
Essential 8™ Medium
Gibco™ Advanced DMEM/F-12
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ Advanced DMEMF/12 is used in cell culture processing applications.
Gibco™ Ham's F-12 Nutrient Mix
Designed for serum-free, single-cell plating of Chinese hamster ovary (CHO) cells
Gibco™ RPMI 1640 Medium (ATCC modification)
Originally developed to culture human leukemic cells in suspension and as a monolayer
Gibco™ Neurobasal™-A Medium
Neurobasal™-A Medium
Gibco™ DMEM, high glucose, pyruvate
Used as basal medium for supporting the growth of many different mammalian cells. Gibco™ DMEM, High Glucose, Pyruvate is used in cell culture processing applications.
Corning™ cellgro™ DMEM Liquid Classical Cell Culture Media
De producten ondergaan diverse testen om de functionaliteit en toxiciteit te bepalen met een groeibevorderingsanalyse. Corning™ cellgro™ DMEM vloeibare klassieke celcultuurmedia ondersteunen celcultuurgroei en vervanging van Pyridoxine HCl door Pyridoxal zorgt voor betere stabiliteit dan bij de oorspronkelijke samenstelling.
Cytiva (Formerly GE Healthcare Life Sciences) Ficoll-Paque™ PLUS
Erkende standaard in laboratoria over de hele wereld voor de isolatie van menselijke lymfocyten uit perifeer bloed. GE Healthcare™ Ficoll-Paque™ PLUS-media handhaaft de levensvatbaarheid en een representatieve distributie van B- en T-lymfocyten.
Gibco™ RPMI 1640 Medium, no glutamine, no phenol red
Animal origin-free
Gibco™ Expi293™ Expression Medium
Expi293™ Expression Medium
Gibco™ Essential 8™ Flex Medium Kit
Essential 8™ Flex is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) without the need for weekend feeding. Gibco™ Essential 8™ Flex Medium Kit is provided in a convenient two-component kit (500mL basal medium and 10mL supplement).
Gibco™ Leibovitz's L-15 Medium, no phenol red
Supports growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue