Reagentia voor chromatografie en massaspectometrie
Thermo Scientific™ Methoxamine (MOX) reagens
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent.
Thermo Scientific™ BSTFA + 1% TMCS-reagens voor silylering
Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1% TMCS Silylation Reagent is derivatizing compounds.
Thermo Scientific™ Pierce™ dimethylsulfoxide (DMSO)
A sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.
Triethylamine solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
CAS: 121-44-8 Moleculaire formule: C6H15N Molecular Weight (g/mol): 101.193 InChI Key: ZMANZCXQSJIPKH-UHFFFAOYSA-N PubChem CID: 8471 ChEBI: CHEBI:35026 IUPAC Name: N,N-diethylethanamine SMILES: CCN(CC)CC
Propionate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
Moleculaire formule: C2H5COO-
Succinate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
Moleculaire formule: OOC(CH2)2COO2-
Bromide Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Maleate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
Moleculaire formule: C2H2(COO)22-
Cyanide solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
Moleculaire formule: Cn-
Thermo Scientific™ Pierce™-trifluorazijnzuur (TFA)
Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.
Thiosulphate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
Moleculaire formule: S2O32
Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) reagens
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,
Nitrate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
CAS: 14797-55-8 Moleculaire formule: NO3 Molecular Weight (g/mol): 62.01 InChI Key: NHNBFGGVMKEFGY-UHFFFAOYSA-N IUPAC Name: nitrate SMILES: [O-][N+]([O-])=O
Multi 7 ions, 10mg/L of each Component, for Ion Chromatography, H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Thermo Scientific™ Pierce™ water, LC/MS kwaliteit
Thermo Scientific™ Pierce Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC
Thermo Scientific™ Pierce™ 0.1% Formic Acid (v/v) in Water, LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents
Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).
Ammonium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
Moleculaire formule: NH4+
Fluoride, Ion chromatography standard solution, Specpure™ F|- 1000μg/mL, Thermo Scientific™
CAS: 7681-49-4 Moleculaire formule: FNa Molecular Weight (g/mol): 41.988 MDL-nummer: MFCD00144315 InChI Key: PUZPDOWCWNUUKD-UHFFFAOYSA-M PubChem CID: 5235 ChEBI: CHEBI:28741 IUPAC Name: sodium;fluoride SMILES: [F-].[Na+]
Thermo Scientific™ BSTFA Silylation Reagent
Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI).
Acetate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
CAS: 127-09-3 Moleculaire formule: C2H3NaO2 Molecular Weight (g/mol): 82.03 MDL-nummer: MFCD00012459 InChI Key: VMHLLURERBWHNL-UHFFFAOYSA-M PubChem CID: 517045 ChEBI: CHEBI:32954 IUPAC Name: sodium;acetate SMILES: [Na+].CC([O-])=O
Thermo Scientific™ Pierce™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Thermo Scientific™ Pierce™ Dimethylformamide (DMF), Sequencing grade
Perform liquid chromatography (HPLC) and mass spectrometry (LC-MS) applications with this 99.5%-pure DMF solvent (MW 73.09, CAS 68-12-2) with low UV-absorptivity.
Triethanolamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
CAS: 9005-65-6 Moleculaire formule: HN(C2H4OH)3+
Thermo Scientific™ Pierce™-trifluorazijnzuur (TFA)
Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.
Thermo Scientific™ TMSI reagens voor silylering
Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent.
Monoethanolamine solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
CAS: 141-43-5 Moleculaire formule: C2H7NO Molecular Weight (g/mol): 61.08 MDL-nummer: MFCD00008183 InChI Key: HZAXFHJVJLSVMW-UHFFFAOYSA-N PubChem CID: 700 ChEBI: CHEBI:16000 IUPAC Name: 2-aminoethanol SMILES: NCCO