Water- en afvalwatermonsternemers
Cole-Parmer™ Wildco 1510-C20 Kemmerer Water Sampler
Cole-Parmer™ Wildco 1510-C20 Kemmerer Water Sampler
Sample at specific depths between 3 and 600 feet. Cole-Parmer™ Wildco 1510-C20 Kemmerer Water Sampler features an acrylic bottle with silicone end seals.
Cole-Parmer™ Polyester Cable
Cole-Parmer™ Polyester Cable
Use this solid braided line with sampling equipment that weighs less than 75 lb (34 kg)
Cole-Parmer™ Turbiditeitsschijf
Cole-Parmer™ Turbiditeitsschijf
Use this disk to determine turbidity or degree of visibility in natural water supplies. Cole-Parmer™ LaMotte™ Secchi Turbidity Disk features braided, stretch resistant lines marked at half and full meter intervals up to 20 meters.
Cole-Parmer™ Wildco™ Horizontal Alpha Water Sampler
Cole-Parmer™ Wildco™ Horizontal Alpha Water Sampler
Suitable for use in oceans, deep lakes and corrosive waters
Cole-Parmer™ Wildco™ Kemmerer Water Sampler
Cole-Parmer™ Wildco™ Kemmerer Water Sampler
Sample at specific depths between 3 and 600 feet.
Cole-Parmer™ Disposable Polypropylene Griffin Low-form Beakers
Cole-Parmer™ Disposable Polypropylene Griffin Low-form Beakers
Molded-in stacking ribs prevent beakers from sticking together when stacked
Thermo Scientific™ Nalgene™ Storm Water Samplers and Mounting Kit
Thermo Scientific™ Nalgene™ Storm Water Samplers and Mounting Kit
A convenient and affordable device for collecting quarterly first flush storm water grab samples in compliance with EPA sampling requirements.
DWK Life Sciences Wheaton™ Sub Surface Grab Sampler II
DWK Life Sciences Wheaton™ Sub Surface Grab Sampler II
For accessing fluid flow in deep manholes and hard-to-reach subsurface sampling areas
DWK Life Sciences Wheaton™ Sub-Surface Grab Sampler™ I System
DWK Life Sciences Wheaton™ Sub-Surface Grab Sampler™ I System
For convenient gathering of subsurface water samples for pollution testing from spillways, boats or docks without entering the water. Assists in conforming to procedures as described in U.S. EPA Handbook for Monitoring Industrial Wastewater.
Buerkle™ Stainless Steel Beaker, Extended for Telescopic Rod
Buerkle™ Stainless Steel Beaker, Extended for Telescopic Rod
Stainless steel sampling dipper with a completely closed, extended handle.
Buerkle™ Beker voor monstername
Buerkle™ Beker voor monstername
Beaker with scraper, contents 1000 ml. Ideal for powder, granules, pastes, mud.
Buerkle™ Pendulum Beaker PP for Buerkle™ Telescopic Rod
Buerkle™ Pendulum Beaker PP for Buerkle™ Telescopic Rod
This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (150 mm dia.). Designed for use with the Buerkle™ Telescopic Rod.
Buerkle™ Aqua Sampler
Buerkle™ Aqua Sampler
Sampler for ground water samples from wells, pits and other sources.
Buerkle™ ViscoSampler
Buerkle™ ViscoSampler
ViscoSampler with powerful suction for viscous media. The media is drawn up and discharged using the suction flask. Specially developed for viscous substances such as slurries, silt, wet clay and soil samples, soil sediments, oils, emulsions, creams etc.
Buerkle™ Chemistry Scoop
Buerkle™ Chemistry Scoop
Scoop for aggressive liquids, complete with strong handle, entirely made of polypropylene.
Buerkle™ Beker voor monstername
Buerkle™ Beker voor monstername
Schep vloeistoffen voor bemonstering met de combinatiebeker en lange handgreep. Buerkle™-bekers voor monstername zijn verkrijgbaar voor het laboratorium, de industrie en waterbehandeling.
Buerkle™ Straight Suction Tube for Buerkle™ ProfiSampler
Buerkle™ Straight Suction Tube for Buerkle™ ProfiSampler
These disposable tubes can be immersed absolutely vertically into the container.
Sampling Systems™ Deep Water Sampler
Sampling Systems™ Deep Water Sampler
Operate this sampler with ease by simply lowering the sampler to the required depth and then pulling it out. Sampling Systems™ Deep Water Sampler is ideal for sampling at large depths.
Buerkle™ Dipping Vessel
Buerkle™ Dipping Vessel
Dipping Vessel for taking all-layer samples from tanks, tank wagons, water courses, water purification plants.
Buerkle™ Extension Tube for Aqua Sampler
Buerkle™ Extension Tube for Aqua Sampler
Aids in obtaining ground water samples from wells, pits and other sources.
Buerkle™ Vampire Sampler
Buerkle™ Vampire Sampler
Transportable, hose pump for liquids, independent of mains power. Ideal for sampling from barrels, tanks, wells, water separators and sewage and purification systems.Vampire is packed in a handy transport case with: drive unit, two rechargeable batteries, battery charger, LED lamp, bottle holder, bottle LDPE 250 ml, hose pump and 5 m hose made of PE.
Buerkle™ Polypropyleen flessenhouder
Buerkle™ Polypropyleen flessenhouder
Kan alle soorten kunststof, glazen of metalen monsterflessen bevatten tot een diameter van 88 mm en een diepte van 6 m. Buerkle™ polypropyleen flessenhouder maakt gebruik van een uitschuifbare stang die is ontworpen voor bemonstering in de industrie en waterbehandeling.
  spinner