Microbiële identificatie
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Thermo Scientific™ ProSpecT™ Giardia/Cryptosporidium Microplate Assay
Thermo Scientific™ ProSpecT™ Giardia/Cryptosporidium Microplate Assay
Rapidly detect Giardia and Cryptosporidium specific antigen from human fecal samples.
Thermo Scientific™ Streptex Latex Group A
Thermo Scientific™ Streptex Latex Group A
Streptex Latex Group A is a supplementary reagent that is used with the streptococcal grouping system Streptex kit.
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using this test kit.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella H
Used for identification of flagellar H antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Oxoid™ X-factor schijfjes
Thermo Scientific™ Oxoid™ X-factor schijfjes
Perform identification of Haemophilus spp. with Thermo Scientific™ Oxoid X Factor Discs.
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Thermo Scientific™ Remel™ Agglutinating Sera, Neisseria meningitidis
Used for serological identification of N. meningitidis by rapid slide agglutination and CIE procedures.
Thermo Scientific™ Remel™ Wellcolex™ E. coli O157
Thermo Scientific™ Remel™ Wellcolex™ E. coli O157
Remel Wellcolex and E. coli O157 is a rapid latex agglutination test used for presumptive identification of E. coli O157 isolates.
Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit
Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit
Eliminates need for incubation and provides a faster strep grouping solution than more traditional enzyme extraction.
Thermo Scientific™ Streptex Positive Control
Thermo Scientific™ Streptex Positive Control
Streptex Positive Control is a polyvalent positive control that is used with the streptococcal Grouping system Streptex kit.
Thermo Scientific™ Oxoid™ Microbact™ Staphylococcal 12S Kit
Thermo Scientific™ Oxoid™ Microbact™ Staphylococcal 12S Kit
Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staphylococcal 12S Kit.
Thermo Scientific™ Staphaurex™ Plus-latexagglutinatietest
Thermo Scientific™ Staphaurex™ Plus-latexagglutinatietest
Identify staphylococci which possess clumping factor, protein A, and/or surface antigens characteristic of MRSA and MSSA strains.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ DrySpot™ E. coli O157 Latex Agglutination Test
Thermo Scientific™ DrySpot™ E. coli O157 Latex Agglutination Test
Rapidly confirm E. coli serogroup O157 from colonies cultured on plated media by latex agglutination.
Thermo Scientific™ Streptex Latex Group D
Thermo Scientific™ Streptex Latex Group D
Streptex Latex Group D is a supplementary reagent that is used with the streptococcal grouping system Streptex kit.
Thermo Scientific™ Streptex Latex Group G
Thermo Scientific™ Streptex Latex Group G
Streptex Latex Group G is a supplementary reagent that is used with the streptococcal grouping system Streptex kit.
Thermo Scientific™ Remel™ Coagulase Plasma
Thermo Scientific™ Remel™ Coagulase Plasma
Thermo Scientific™ Remel Coagulase Plasma is used for detection of coagulase enzyme in staphylococci.
Thermo Scientific™ Oxoid™ IMAGEN™ Chlamydia Positive Control Slide
Thermo Scientific™ Oxoid™ IMAGEN™ Chlamydia Positive Control Slide
Oxoid IMAGEN Chlamydia Positive Control Slide contains acetone-fixed human epithelial cells infected with Chlamydia.
Thermo Scientific™ DrySpot™ Legionella Pneumophila Serogroup 2-14 Latex Agglutination Test
Thermo Scientific™ DrySpot™ Legionella Pneumophila Serogroup 2-14 Latex Agglutination Test
Rapidly confirm Legionella pneumophila serogroups 2-14 from culture by latex agglutination.
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O/Vi
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O/Vi
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Thermo Scientific™ Neisseria meningitidis  Agglutinating Sera
Thermo Scientific™ Neisseria meningitidis Agglutinating Sera
Meningococcus Agglutinating Sera are used in the qualitative serological identification of Neisseria meningitidis Groups A, B, C, D, X, Y, Z, W135, A-D and X-Z + W135.
Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay
Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay
Rapidly detect E. coli Shiga Toxin 1 and 2 from human fecal samples.
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Thermo Scientific™ DrySpot™ Legionella Pneumophila Serogroup 1 Latex Agglutination Test
Thermo Scientific™ DrySpot™ Legionella Pneumophila Serogroup 1 Latex Agglutination Test
Rapidly confirm Legionella pneumophila serogroup 1 from culture by latex agglutination.
Thermo Scientific™ Remel™ BactiCard™ Strep
Thermo Scientific™ Remel™ BactiCard™ Strep
Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.
Thermo Scientific™ Streptex™ Latex Agglutination Test
Thermo Scientific™ Streptex™ Latex Agglutination Test
Isolate and identify a variety of streptococci with a simple enzyme extraction method that detects Lancefield groups A, B, C, D, F and G.
Thermo Scientific™ Oxoid™ X & V Factor Discs
Thermo Scientific™ Oxoid™ X & V Factor Discs
For identification of Haemophilus spp.
Thermo Scientific™ PathoDx Strep Grouping Universal Kit
Thermo Scientific™ PathoDx Strep Grouping Universal Kit
Support the functionality of the Thermo Scientific™ PathoDx™ Strep Grouping Kit with Thermo Scientific™ PathoDx™ Strep Grouping Universal Kit.
Thermo Scientific™ DrySpot™ Pneumo Latex Agglutination Test
Thermo Scientific™ DrySpot™ Pneumo Latex Agglutination Test
Rapidly detect capsular antigen from Streptococcus pneumoniae isolated from culture plates and blood culture by latex agglutination.
  spinner