pH-referentiebuffers
Thermo Scientific™ Orion™ Standard All-in-One™ pH-bufferkit
Thermo Scientific™ Orion™ Standard All-in-One™ pH-bufferkit
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
pH-bufferoplossing voor laboratorium, pH 4.01
pH-bufferoplossing voor laboratorium, pH 4.01
Designed for use in laboratory pH measurement. Reagecon™ Laboratory Buffer Solution, pH 4.01, Red is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability and is supplied with a detailed Certificate of Analysis.
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
Redox-buffer voor het testen van ORP-elektroden
Redox-buffer voor het testen van ORP-elektroden
Used for testing, calibration and verification of redox/ORP sensors. Redox Buffer, 220mV, Reagecon™ is a ready to use solution of iron salts, including potassium ferrocyanide and tripotassium hexacyanoferrate, for use with reactions with a redox potential of 220mV.
pH-bufferoplossing voor laboratorium, pH 7.00
pH-bufferoplossing voor laboratorium, pH 7.00
Designed for use in laboratory pH measurement. Reagecon™ Laboratory Buffer Solution, pH 7.00, Green is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability and is supplied with a detailed Certificate of Analysis.
pH-bufferoplossing voor laboratorium, pH 9.21
pH-bufferoplossing voor laboratorium, pH 9.21
Designed for use in laboratory pH measurement. Reagecon™ Laboratory Buffer Solution, pH 9.21, Blue is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability and is supplied with a detailed Certificate of Analysis.
pH 4.01 Technical Color-Coded Buffer Solution at 25°C, Reagecon
pH 4.01 Technical Color-Coded Buffer Solution at 25°C, Reagecon
Easy-to-use, tested, certified, NIST traceable, and shelf-stable, color-coded buffer solution.
pH 9.00 Buffer Solution at 25°C, Reagecon
pH 9.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Standaard bufferoplossing pH 4.01
Standaard bufferoplossing pH 4.01
With a high accuracy: ±0.02 pH, this standard buffering solution is traceably certified to PTB/NIST and therefore safe for use.
pH 4.00 Phthalate-Free Buffer Solution at 20°C, Reagecon
pH 4.00 Phthalate-Free Buffer Solution at 20°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
pH 7.00 Buffer Solution at 25°C, Reagecon
pH 7.00 Buffer Solution at 25°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Thermo Scientific™ Orion™ pH Buffer Bottles
Thermo Scientific™ Orion™ pH Buffer Bottles
Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.
Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.
Succinate, 0.2M buffer soln., pH 4.5, Thermo Scientific™
Succinate, 0.2M buffer soln., pH 4.5, Thermo Scientific™
CAS: 150-90-3 Moleculaire formule: C4H4Na2O4 Molecular Weight (g/mol): 162.052 MDL-nummer: MFCD00002789 InChI Key: ZDQYSKICYIVCPN-UHFFFAOYSA-L PubChem CID: 9020 ChEBI: CHEBI:63675 IUPAC Name: disodium;butanedioate SMILES: C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+]
Buffer, Precision Reference Standard, pH 10.000 ± 0.005 at 25°C, Ricca Chemical
Buffer, Precision Reference Standard, pH 10.000 ± 0.005 at 25°C, Ricca Chemical
Standardized against NIST-certified pH references. High-Accuracy pH Buffer solutions are for use with meters with 0.001 resolution.
Thermo Scientific™ Sodium bicarbonate, 1M buffer soln., pH 9.0
Thermo Scientific™ Sodium bicarbonate, 1M buffer soln., pH 9.0
CAS: 144-55-8 Moleculaire formule: CHNaO3 Molecular Weight (g/mol): 84.006 MDL-nummer: MFCD00003528 InChI Key: UIIMBOGNXHQVGW-UHFFFAOYSA-M PubChem CID: 516892 ChEBI: CHEBI:32139 IUPAC Name: sodium;hydrogen carbonate SMILES: C(=O)(O)[O-].[Na+]
  spinner