ELISA-kits en -reagentia
Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (20X)
Invitrogen™ NuPAGE™ MOPS SDS Running Buffer (20X)
Made with high-purity reagents and are strictly quality controlled
Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)
Invitrogen™ Novex™ Tris-Glycine SDS Running Buffer (10X)
Formulated for separation of proteins, in their denatured state, on Novex Tris-Glycine gels
Invitrogen™ Dynabeads™ MyOne™ Streptavidin T1
Invitrogen™ Dynabeads™ MyOne™ Streptavidin T1
Dynabeads MyOne Streptavidin T1 are the gold standard for isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets.
Invitrogen™ NuPAGE™ LDS Sample Buffer (4X)
Invitrogen™ NuPAGE™ LDS Sample Buffer (4X)
Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.
Invitrogen™ IL-6 Human Uncoated ELISA Kit with Plates
Invitrogen™ IL-6 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human IL-6 protein levels from samples including serum, plasma, and cell culture supernatants.
Invitrogen™ NuPAGE™ Antioxidant
Invitrogen™ NuPAGE™ Antioxidant
Proprietary reagent that maintains proteins in a reduced state during protein gel electrophoresis
Invitrogen™ NuPAGE™ Tris-Acetate SDS Running Buffer (20X)
Invitrogen™ NuPAGE™ Tris-Acetate SDS Running Buffer (20X)
Formulated for separation of proteins, in their denatured state, on NuPAGE Novex Tris-Acetate gels
Invitrogen™ ELISA Stop Solution
Invitrogen™ ELISA Stop Solution
For use with enzyme assays which employ horseradish peroxidase and TMB substrate solution
Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X)
Invitrogen™ NuPAGE™ MES SDS Running Buffer (20X)
Made with high-purity reagents and are strictly quality controlled
Invitrogen™ NuPAGE™ Sample Reducing Agent (10X)
Invitrogen™ NuPAGE™ Sample Reducing Agent (10X)
Ready-to-use reagent to reduce protein samples for protein gel electrophoresis
Thermo Scientific™ SuperBlock™ (PBS) Blocking Buffer
Thermo Scientific™ SuperBlock™ (PBS) Blocking Buffer
Block membranes, plates and tissue samples with these optimized solutions containing a protein that provides fast and effective blocking for Western blot, ELISA and IHC.
Thermo Scientific™ Pierce™ TMB-substraten
Thermo Scientific™ Pierce™ TMB-substraten
Horseradish peroxidase (HRP) substrates for ELISA that change from blue to yellow upon addition of a sulfuric or phosphoric acid stop solution.
Thermo Scientific™ Pierce™ TMB-substraten
Thermo Scientific™ Pierce™ TMB-substraten
Horseradish peroxidase (HRP) substrates for ELISA that change from blue to yellow upon addition of a sulfuric or phosphoric acid stop solution.
Invitrogen™ Dynabeads™ MyOne™ Streptavidin C1
Invitrogen™ Dynabeads™ MyOne™ Streptavidin C1
Exceptional standard for isolation and handling of biotinylated nucleic acids, antibodies, or other biotinylated ligands and targets
Invitrogen™ Mouse TNF alpha ELISA Kits
Invitrogen™ Mouse TNF alpha ELISA Kits
Measure levels of mouse tumor necrosis factor alpha (TNFa) in mouse serum and cell culture supernatant samples using 96-well plates.
IBA Lifesciences Avidin
IBA Lifesciences Avidin
Avidin can efficiently block of biotin and biotinlyated proteins.
Thermo Scientific™ Pierce™ StartingBlock™-blokkeringsbuffers
Thermo Scientific™ Pierce™ StartingBlock™-blokkeringsbuffers
Block membranes, plates and other samples with these ready-to-use TBS formulations of a special protein blocker for exceptional nonspecific blocking and broad compatibility.
Invitrogen™ TGF beta-1 Human/Mouse Uncoated ELISA Kit with Plates
Invitrogen™ TGF beta-1 Human/Mouse Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of human or mouse TGF beta 1 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range of 8-1000pg/mL.
Invitrogen™ Bolt™ Antioxidant
Invitrogen™ Bolt™ Antioxidant
A proprietary reagent that maintains proteins in a reduced state during protein gel electrophoresis
Thermo Scientific™ Pierce™ SEA BLOCK™ blokkeringsbuffer
Thermo Scientific™ Pierce™ SEA BLOCK™ blokkeringsbuffer
Block mammalian tissue samples for immunohistochemistry (IHC) and samples for other methods with steelhead salmon serum in PBS.
Molecular Probes™ Streptavidin, Alexa Fluor™ 594 Conjugate
Molecular Probes™ Streptavidin, Alexa Fluor™ 594 Conjugate
Streptavidin, Alexa Fluor™ 594 conjugate
Thermo Scientific™ Pierce™ Human IFN alpha ELISA Kits (Multi-Subtype)
Thermo Scientific™ Pierce™ Human IFN alpha ELISA Kits (Multi-Subtype)
Measure levels of 14 identified subtypes of human interferon alpha (IFNa) in human cell culture media supernatant samples using 96-well plates.
Thermo Scientific™ 6X DNA-laadkleurstof
Thermo Scientific™ 6X DNA-laadkleurstof
Prepare DNA markers and samples for loading on agarose or polyacrylamide gels with Thermo Scientific™ 6X DNA Loading Dye.
Thermo Scientific™ QuantaBlu™ fluorogeen peroxidasesubstraat
Thermo Scientific™ QuantaBlu™ fluorogeen peroxidasesubstraat
Conduct stopped, non-stopped and kinetic assays using this soluble fluorogenic (chemifluorescent) peroxidase substrate for ELISA.
  spinner