Reagentia, membranen en toebehoren voor western blotting
Invitrogen™ PVDF/Filter Paper Sandwich, 0.2 µm, 8.3 x 7.3 cm
Pre-cut and pre-assembled—ready for protein blotting/transfer
Controlestof voor humaan antiserum, Thermo Scientific™
Human Control
Thermo Scientific™ Pierce™ ECL-substraat voor western blotting
Value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.
Cytiva Amersham™ AlkPhos Direct™ Labeling Modules
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes. AlkPhos Direct Labeling Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase.
Invitrogen™ iBlot™ 2 Gel Transfer Device
Performs Western blotting transfer simply, efficiently, and reliably, within seven minutes and without the need for liquid buffers
Thermo Scientific™ SuperSignal™ West Pico PLUS Chemiluminescent Substrate
An enhanced chemiluminescent (ECL) horseradish peroxidase (HRP) substrate that enables picogram- to high femtogram-level protein detection by western blot analysis
Thermo Scientific™ SuperSignal™ West Femto chemiluminescent substraat
Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.
Invitrogen™ CM-H2DCFDA (General Oxidative Stress Indicator)
General oxidative stress indicator
Invitrogen™ iBlot™ 2 Transfer Stacks, PVDF, regular size
Consumable stacks with integrated PVDF transfer membranes for dry blotting of proteins
Cytiva Amersham™ CDP-Star detectiereagentia
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic system
- High flowrates and good scale-up potential
Thermo Scientific™ Pierce™ Protein-Free (PBS) Blocking Buffer
Block membranes, plates and other samples with these ready-to-use PBS and TBS formulations of a non-protein compound to yield extremely low background.
Thermo Scientific™ Pierce™ streptavidine HRP-conjugaat, hoge gevoeligheid
Amplify signal in Western blot, ELISA or other biotin-based detection methods with this exclusive, high signal-to-noise, peroxidase-conjugated, ready-to-use streptavidin.
Thermo Scientific™ Pierce™ recombinant eiwit A/G en conjugaten
Detect antibodies, especially IgG isotypes, from many different species hosts with this purified, alkaline phosphatase- or peroxidase-conjugated recombinant Protein A/G.
Thermo Scientific™ CL-XPosure™-film (röntgenfilm)
Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.
Thermo Scientific™ Blocker™ Casein in PBS
Block membranes, plates and other samples with these ready-to-use PBS solutions of purified casein for Western blot, ELISA, and other detection methods.
Cytiva Amersham™ AlkPhos Direct™ ECF Detection Module
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Thermo Scientific™ Pierce™ PVDF-membranen
Enable high protein and nucleic acid binding capacity with sheets and rolls of high-quality polyvinylidene difluoride (PVDF) membrane.
Thermo Scientific™ Blocker™ BLOTTO blokkeringsbuffer
Block membranes, plates and other samples with this ready-to-use 5% solution of nonfat powdered milk in TBS for Western blot, ELISA and other detection methods.
Thermo Scientific™ Pierce™ Avidin
Perform Western blot, ELISA or other biotin-based detection methods using these purified and HRP-, AP- or FITC-labeled avidin molecules.
Thermo Scientific™ CL-XPosure™-film (röntgenfilm)
Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.
Invitrogen™ XCell SureLock™ Mini-Cell and XCell II™ Blot Module
Quickly and easily run and blot Novex minigels
Invitrogen™ iBlot™ 2 Transfer Stacks, nitrocellulose, regular size
Used to transfer proteins with the iBlot 2 Gel Transfer Device
Thermo Scientific™ Pierce™ Clear Milk Blocking Buffer (10X)
Thermo Scientific™ Pierce Clear Milk Blocking Buffer is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.
Cytiva Amersham™ Protran™ NC-nitrocellulosemembranen: Rollen
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Cytiva Elektroden en papier voor NovaBlot™
These Electrodes and Paper for NovaBlot™ are accessory items for NovaBlot™ Kit, the Multiphor™ II Electrophoresis System.
Thermo Scientific™ Restore™ Fluorescent Western Blot Stripping Buffer
Quickly and gently remove primary and near-infrared (IR) dye-labeled secondary antibodies from Western blots.
Invitrogen™ HPF (Hydroxyl Radical and Peroxynitrite Sensor)
Two indicators that together can selectively detect hypochlorite anion
Invitrogen™ Novex™ ECL Chemiluminescent Substrate Reagent Kit
Chemiluminescent substrate used for chemiluminescence-based immunodetection of horse radish peroxidase (HRP) on western or dot blot membranes
Thermo Scientific™ Pierce™ streptavidine poly-HRP conjugaat
Amplify signal and improve detection of biotinylated antibodies for IHC and other methods using this streptavidin conjugated with HRP polymers.