Chromatography Spare Parts
Cytiva PEEK Stop Plug
Cytiva PEEK Stop Plug
Enables connection of HiTrap columns to a syringe, laboratory pump or chromatography system. Cytiva PEEK Stop Plugs are used to seal the top of a chromatography column, e.g. HiTrap™ prepacked columns. Fingertight or standard 1/16 in. or standard 5/16 in. male stop plugs available.
Cytiva Silicone Tubing
Cytiva Silicone Tubing
Silicone tubing for Peristaltic Pump P-1.  Cytiva Silicone Tubing with i.d. 3.1 mm.
Cytiva ETFE Ferrule
Cytiva ETFE Ferrule
Ferrules used for chromatography tubing. Cytiva Lifescience™ ETFE Ferrule is made from ethylenetetra fluoroethylene and comes with different outside diameters.
SCAT™ Fittings for HPLC Systems
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
Vici Valco Instruments™ Stainless Steel Tubing Cutter
Vici Valco Instruments™ Stainless Steel Tubing Cutter
Achieve smooth and uniform cutting of stainless steel tubing with this cutting wheel type tubing cutter. Vici Valco Instruments™ Stainless Steel Tubing Cutter is ideal for cutting 1/16 in. and 1/8 in. SS tubing.
Thermo Scientific™ General Gas Chromatography Tubing and Tools
Thermo Scientific™ General Gas Chromatography Tubing and Tools
Use Thermo Scientific™ General Gas Chromatography Tubing and Tools to maintain and optimize your GC instrument.
Cytiva PEEK Tubing
Cytiva PEEK Tubing
PEEK tubing comes in a variety of internal diameter sizes.  Cytiva Lifescience™ PEEK Tubing is an important chromatography accessory.
Cytiva PEEK Tubing Kit, I.D. 0.50mm
Cytiva PEEK Tubing Kit, I.D. 0.50mm
For use with ÄKTA pure 150
Cytiva Polypropylene On-line Filter Kit
Cytiva Polypropylene On-line Filter Kit
Used for AKTA chromatography systems.  Cytiva Polypropylene On-line Filter Kits are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules.
Cytiva PEEK Finger Tight Connector
Cytiva PEEK Finger Tight Connector
Use With Tubing o.d. 1/16 in. Cytiva PEEK Finger Tight Connector provides the tightest connections.
SGE™ SilFlow™ GC 3 Port Splitter Kits
SGE™ SilFlow™ GC 3 Port Splitter Kits
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
Thermo Scientific™ Terry Tool Tubing Cutters
Thermo Scientific™ Terry Tool Tubing Cutters
Produce clean, 90° cuts of 1/16 in. and 1/8 in. stainless-steel tubing used in your HPLC system with Thermo Scientific™ Terry Tool Tubing Cutters.
Thermo Scientific™ GC Tool Kit for Agilent Instruments
Thermo Scientific™ GC Tool Kit for Agilent Instruments
Thermo Scientific™ GC Tool Kit for Agilent Instruments includes common consumable supplies and tools.
Cytiva PEEK Connector 1/16 in. Luer
Cytiva PEEK Connector 1/16 in. Luer
Set of two (2) polyetheretherketone 1/16 in. Male/Luer Female connectors for connection of a syringe to the top of a chromatography column in the ÄKTA Chromatography System.
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Thermo Scientific™ Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Thermo Scientific™ Gas Chromatography GLD Pro Gas Leak Detector and GFM Pro Electronic Flowmeter Accessories
Probe adaptor and carrying case for use with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Thermo Scientific™ PEEK Capillary Tubing
Thermo Scientific™ PEEK Capillary Tubing
Use these sample loops for your Thermo Scientific™ Rheodyne™ HPLC sample injectors.
Thermo Scientific™ Septa
Thermo Scientific™ Septa
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Cytiva Borosilicate Glass XK Chromatography Tubes
Cytiva Borosilicate Glass XK Chromatography Tubes
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules.
Cytiva ETFE Capillary Tubing
Cytiva ETFE Capillary Tubing
For use with ÄKTA chromatography systems. Cytiva ETFE Capilary Tubing is available in a range of sizes.
Cytiva Borosilicate Glass Superloops
Cytiva Borosilicate Glass Superloops
Use for the introduction of large sample volumes into a pressurised fluid LC system.  Cytiva Lifescience™ Borosilicate Glass Superloops work in conjunction with 7-port valves. 
  spinner