Learn More
Diméthylphénylsilanol, 97 %
49.90€ - 162.00€
Identifiants chimiques
CAS | 5272-18-4 |
---|---|
Formule moléculaire | C8H12OSi |
Poids moléculaire (g/mol) | 152.27 |
Numéro MDL | MFCD08276879 |
Clé InChI | FDTBETCIPGWBHK-UHFFFAOYSA-N |
CID PubChem | 78929 |
Nom IUPAC | dimethyl(phenyl)silanol |
SMILES | C[Si](C)(O)C1=CC=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
5272-18-4 | |
152.27 | |
FDTBETCIPGWBHK-UHFFFAOYSA-N | |
dimethyl(phenyl)silanol |
C8H12OSi | |
MFCD08276879 | |
78929 | |
C[Si](C)(O)C1=CC=CC=C1 |
Spécification
60°C to 62°C (0.5mmHg) | |
5272-18-4 | |
MFCD08276879 | |
FDTBETCIPGWBHK-UHFFFAOYSA-N | |
dimethyl(phenyl)silanol | |
78929 | |
97% |
Dimethylphenylsilanol | |
C8H12OSi | |
dimethylphenylsilanol, phenyldimethylsilanol, silanol, dimethylphenyl, silanol, 1,1-dimethyl-1-phenyl, dimethyl phenyl silanol, acmc-1avoz, amtsi002, silanol,1,1-dimethyl-1-phenyl | |
C[Si](C)(O)C1=CC=CC=C1 | |
152.27 | |
152.27 |
Sécurité et traitement
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures