Learn More
Sphères en cuivre aluminium, alliage 2017, 1,59 mm (0,063 er) de diamètre, Thermo Scientific™
132.00€ - 393.00€
Spécification
Nom chimique ou matériau | Aluminum Copper spheres, Alloy 2017 |
---|---|
Note relative au nom | 1.59mm (0.063 in.) dia. |
Moy. Mol. Poids ou mol. Plage de poids | Al:Cu:Mn:Mg; 94.8:4:0.7:0.5 wt% |
Numéro MDL | MFCD00213972 |
Odeur | Odorless |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Aluminum Copper spheres, Alloy 2017 | |
Al:Cu:Mn:Mg; 94.8:4:0.7:0.5 wt% | |
Odorless |
1.59mm (0.063 in.) dia. | |
MFCD00213972 |
Sécurité et traitement
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures