Learn More
Alfa Aesar™ 3-Bromothiophene-2-sulfonyl chloride, 90+%
82.50€ - 221.00€
Identifiants chimiques
CAS | 170727-02-3 |
---|---|
Formule moléculaire | C4H2BrClO2S2 |
Poids moléculaire (g/mol) | 261.532 |
Numéro MDL | MFCD08059484 |
Clé InChI | OPLMNCCLQDVCTP-UHFFFAOYSA-N |
Synonyme | 3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo |
CID PubChem | 13675598 |
Nom IUPAC | 3-bromothiophene-2-sulfonyl chloride |
SMILES | C1=CSC(=C1Br)S(=O)(=O)Cl |
Description
Identifiants chimiques
170727-02-3 | |
261.532 | |
OPLMNCCLQDVCTP-UHFFFAOYSA-N | |
13675598 | |
C1=CSC(=C1Br)S(=O)(=O)Cl |
C4H2BrClO2S2 | |
MFCD08059484 | |
3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo | |
3-bromothiophene-2-sulfonyl chloride |
Spécification
3-Bromothiophene-2-sulfonyl chloride | |
39°C to 41°C | |
MFCD08059484 | |
3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo | |
C1=CSC(=C1Br)S(=O)(=O)Cl | |
261.532 | |
261.55 |
170727-02-3 | |
C4H2BrClO2S2 | |
UN3261 | |
OPLMNCCLQDVCTP-UHFFFAOYSA-N | |
3-bromothiophene-2-sulfonyl chloride | |
13675598 | |
≥90% |
Sécurité et traitement
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.