Learn More
Méthacrylate de 2,2,3,3-tétrafluoropropyle, 97 %, stab. avec 50 ppm de 4-méthoxyphénol, Thermo Scientific Chemicals
26.70€ - 99.30€
Identifiants chimiques
CAS | 45102-52-1 |
---|---|
Formule moléculaire | C7H8F4O2 |
Poids moléculaire (g/mol) | 200.133 |
Numéro MDL | MFCD00042381 |
Clé InChI | RSVZYSKAPMBSMY-UHFFFAOYSA-N |
Synonyme | 2,2,3,3-tetrafluoropropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester, methacrylic acid 2,2,3,3-tetrafluoropropyl ester, 1h,1h,3h-tetrafluoropropyl methacrylate, 2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol, acmc-1ajz7, ksc489s8d, rsvzyskapmbsmy-uhfffaoysa, 2,2,3,3-tetrafluoroprop-1-yl methacrylat |
CID PubChem | 123515 |
Nom IUPAC | 2,2,3,3-tétrafluoropropyl 2-méthylprop-2-énoate |
SMILES | CC(=C)C(=O)OCC(C(F)F)(F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
45102-52-1 | |
200.133 | |
RSVZYSKAPMBSMY-UHFFFAOYSA-N | |
123515 | |
CC(=C)C(=O)OCC(C(F)F)(F)F |
C7H8F4O2 | |
MFCD00042381 | |
2,2,3,3-tetrafluoropropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester, methacrylic acid 2,2,3,3-tetrafluoropropyl ester, 1h,1h,3h-tetrafluoropropyl methacrylate, 2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol, acmc-1ajz7, ksc489s8d, rsvzyskapmbsmy-uhfffaoysa, 2,2,3,3-tetrafluoroprop-1-yl methacrylat | |
2,2,3,3-tétrafluoropropyl 2-méthylprop-2-énoate |
Spécification
2,2,3,3-Tetrafluoropropyl methacrylate | |
45102-52-1 | |
146°C to 149°C | |
C7H8F4O2 | |
MFCD00042381 | |
3588831 | |
RSVZYSKAPMBSMY-UHFFFAOYSA-N | |
2,2,3,3-tétrafluoropropyl 2-méthylprop-2-énoate | |
123515 | |
97% |
stabilized with 50ppm 4-methoxyphenol | |
1.254 | |
50°C (122°F) | |
1.375 | |
UN3272 | |
2,2,3,3-tetrafluoropropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester, methacrylic acid 2,2,3,3-tetrafluoropropyl ester, 1h,1h,3h-tetrafluoropropyl methacrylate, 2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol, acmc-1ajz7, ksc489s8d, rsvzyskapmbsmy-uhfffaoysa, 2,2,3,3-tetrafluoroprop-1-yl methacrylat | |
CC(=C)C(=O)OCC(C(F)F)(F)F | |
200.133 | |
200.13 |
Sécurité et traitement
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H226-H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
missing translation for 'einecsNumber' : 256-189-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only