missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ester pinacol d’acide 1-dodécylboronique, 98 %
45.41€ - 486.00€
Identifiants chimiques
CAS | 177035-82-4 |
---|---|
Formule moléculaire | C18H37BO2 |
Poids moléculaire (g/mol) | 296.302 |
Numéro MDL | MFCD12546195 |
Clé InChI | WHOMEQHZQLKAPC-UHFFFAOYSA-N |
Synonyme | 1-dodecylboronic acid pinacol ester, 2-n-dodecyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane |
CID PubChem | 10590017 |
Nom IUPAC | 2-dodécyl-4,4,5,5-tétraméthyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCCCC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
177035-82-4 | |
296.302 | |
WHOMEQHZQLKAPC-UHFFFAOYSA-N | |
10590017 | |
B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCCCC |
C18H37BO2 | |
MFCD12546195 | |
1-dodecylboronic acid pinacol ester, 2-n-dodecyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane | |
2-dodécyl-4,4,5,5-tétraméthyl-1,3,2-dioxaborolane |
Spécification
1-Dodecylboronic acid pinacol ester | |
123°C (0.2 mmHg) | |
1.441 | |
1-dodecylboronic acid pinacol ester, 2-n-dodecyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane | |
B1(OC(C(O1)(C)C)(C)C)CCCCCCCCCCCC | |
296.302 | |
296.31 |
177035-82-4 | |
C18H37BO2 | |
MFCD12546195 | |
WHOMEQHZQLKAPC-UHFFFAOYSA-N | |
2-dodécyl-4,4,5,5-tétraméthyl-1,3,2-dioxaborolane | |
10590017 | |
98% |
Sécurité et traitement
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only