thermo-scientific-spectroscopy-solutions-banner thermo-scientific-spectroscopy-solutions-banner-m
NMR
nuclear-magnetic-resonance-icon
Nuclear Magnetic Resonance (NMR)
Designed to be accessible and affordable, Thermo Scientific™ picoSpin™ nuclear magnetic resonance (NMR) products provide the analytical power of NMR in a desktop solution, ideal for teaching and industrial applications.
Thermo Scientific picoSpin 45 NMR Spectrometer
True proton NMR spectroscopy without the expense or complexity of traditional systems. Samples are simply injected into the front panel of the instrument.
The temperature-controlled permanent magnet requires no liquid cryogens. Magnet temperature: 8°C above ambient temperature to 45°C.
Buy picoSpin 45 NMR Spectrometer
Thermo Scientific picoSpin 45 NMR Spectrometer - 19Fluorine
  • 19Fluorine NMR spectroscopy without the expense or complexity of traditional systems. Samples are simply injected into the front panel of the instrument
  • The temperature-controlled permanent magnet requires no liquid cryogens
  • Magnet temperature: 8°C above ambient temperature to 45°C
Buy the picoSpin 45 NMR Spectrometer - 19 Fluorine
Microvolume UV-Vis
microvolume-uv-vis-icon
Microvolume UV-Vis
Trust Thermo Scientific NanoDrop instruments to reliably quantify 1µL samples using patented sample-retention technology. Measure DNA, RNA and proteins in five seconds – pipette, measure and wipe clean.
Thermo Scientific NanoDrop Lite Spectrophotometer
Portable and compact for basic DNA, RNA and protein measurements.
Buy the Nanodrop Lite
Buy the Nanodrop Lite and printer package
Thermo Scientific NanoDrop 2000 Spectrophotometer
Obtain full-spectrum DNA, RNA and protein measurements with incredible reproducibility.
Buy the NanoDrop 2000
Buy the Nanodrop 2000 and laptop computer package
Thermo Scientific NanoDrop 2000c Spectrophotometer
Perform full-spectrum microvolume or cuvette measurements of nucleic acid and protein.
Buy the NanoDrop 2000c
Buy the Nanodrop 2000c and laptop computer package
Thermo Scientific NanoDrop 8000 Spectrophotometer
Obtain full-spectral measurements of eight samples simultaneously using an 8-channel pipette.
Buy the NanoDrop 8000 
Buy the Nanodrop 8000 and laptop computer package
Spectrophotometers
Spectrophotometers
Select the UV-visible or visible spectrophotometer for your teaching, quality assurance or research needs. Get answers at the speed of light with these reliable tools you can use every day.
Nicolet iS5 FT-IR Spectrometer
The Nicolet IS5 FT-IR Spectrometer sets a new standard for IR Spectrometers, making identification, quality screening, or teaching easier and more reliable than ever. The Nicolet IS5 also comes with software and a library of known samples to assist in identifying unknown substances.
thermo-scientific-spectronic-200-visible-spectrophotometer
Thermo Scientific SPECTRONIC 200 Visible Spectrophotometer
The modern gold standard for value-priced spectrophotometry in teaching and basic analysis.
Buy the SPECTRONIC 200 Visible Spectrophotometer
thermo-scientific-genesys-20-spectrophotometer
Thermo Scientific GENESYS 20 Visible Spectrophotometer
Compact and reliable measurements in absorbance, transmittance or concentration. Ideal for education and industrial labs.
Buy the GENESYS 20
Buy the GENESYS 20 with built-in printer
thermo-scientific-genesys-10s-spectrophotometer
Thermo Scientific GENESYS 10S UV-Vis Spectrophotometer
Accurate performance-proven instrument with fast scanning and three-year xenon lamp warranty for low maintenance.
Buy the GENESYS 10S UV-Vis
Buy the GENESYS 10S UV-Vis with internal printer
thermo-scientific-genesys-10s-spectrophotometer
Thermo Scientific GENESYS 10S Visible Spectrophotometer
Ideal for teaching basic spectroscopy with a USB memory device for sharing data.
Buy the GENESYS 10S Visible
Buy the GENESYS 10S Visible with internal printer
thermo-scientific-biomate-3s-spectrophotometer
Thermo Scientific BioMate 3S Spectrophotometer
Automated UV-Vis analysis for life science measurements of DNA , RNA and protein. Follow enzyme kinetics and monitor cell growth.
Buy the BioMate 3S
Buy the BioMate 3S with printer
thermo-scientific-evolution-201-220-spectrophotometers
Thermo Scientific Evolution 201/220 UV-Vis Spectrophotometers
Our most innovative, award-winning spectrophotometers offer double-beam high performance, flexibility and reliability.
Buy the Evolution 201 PC 
Buy the Evolution 201 LC 
Buy the Evolution 220 PC
Buy the Evolution 220 LC
Buy the Evolution SmartSipper Accessory
thermo-scientific-evolution-260-bio-spectrophotometer
Thermo Scientific Evolution 260 Bio UV-Vis Spectrophotometer
Next generation INSIGHT software provides streamlined convenience of nucleic acid and protein analysis methods.
Buy the Evolution 260 PC
Buy the Evolution 260 LC
thermo-scientific-evolution-300-spectrophotometer
Thermo Scientific Evolution 300 UV-Vis Spectrophotometer
No warm-up time needed with a xenon lamp system that matches accessories and variable bandwidths to user assays.
Buy the Evolution 300 PC with VisionSecurity Software
Buy the Evolution 300 PC with VisionPro Software
FT-IR Fluorescence
fluorescence-icon
Fluorescence
Rely on the sensitivity of fluorescence detection to measure DNA, RNA and proteins labeled with fluors for PCR, FRET assays and routine quantitation.
thermo-scientific-nanodrop2000c.png
Thermo Scientific NanoDrop 3300 Fluorospectrometer
Full-spectrum analyses of fluorescence emission (400-750nm) for microliter samples. Measure GFP and DNA-binding fluors (e.g., PicoGreen) without changing filters using high-performance LEDs.
Buy the NanoDrop 3300
Buy the NanoDrop 3300 with laptop computer
New Product: Thermo Scientific NanoDrop One
Learn more about the NanoDrop One Microvolume UV-Vis Spectrophotometer and its Acclaro Sample Intelligence Technology.
Special Offers
Resources
More