missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YME1L1 Mouse anti-Human, Rat, Clone: 2B7E4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66551-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.Spécification
YME1L1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q925S8, Q96TA2 | |
Yme1l1 | |
YME1L1 Fusion Protein Ag2069 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
2B7E4 | |
Unconjugated | |
YME1L1 | |
FTSH, FTSH1, Meg 4, MEG4, PAMP, YME1 like 1 (S. cerevisiae), YME1 like protein 1, YME1L, YME1L1 | |
Mouse | |
Protein G | |
RUO | |
10730, 114217 | |
-20°C | |
Liquid |