missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSX2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25825-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CHX10 encodes a homeobox protein originally described as a retina-specific transcription factor. Mutations in this gene are associated with microphthalmia, cataracts and iris abnormalities.Spécification
VSX2 | |
Polyclonal | |
Unconjugated | |
VSX2 | |
CHX10, Homeobox protein CHX10, HOX10, MCOP2, MCOPCB3, RET1, visual system homeobox 2, VSX2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12677, 338917 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P58304, Q61412 | |
VSX2 | |
VSX2 Fusion Protein Ag23035 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |