missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Rabbit anti-Human, Mouse, Rat, CoraLite 594, Clone: 6K21, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marque: Proteintech CL594-80232-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Vimentin Recombinant Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for IFSpécification
Vimentin | |
Recombinant Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Rabbit | |
20 μL | |
7431 | |
Store at -20°C | |
Liquid |
Immunofluorescence | |
6K21 | |
CoraLite 594 | |
BC000368 | |
Protein A purification | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |