Learn More
UPF1 Mouse anti-Human, Unconjugated, Clone: 3B6B5, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 66898-1-IG-20UL
Code produit. 16869175
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
The UPF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UPF1. This antibody recognizes human, mouse, rat antigen. The UPF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
UPF1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
3B6B5
Unconjugated
20 μL
5976
IgG1