Learn More
UPF1 Mouse anti-Human, Unconjugated, Clone: 3B6B5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66898-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The UPF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UPF1. This antibody recognizes human, mouse, rat antigen. The UPF1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
UPF1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
3B6B5 | |
Unconjugated | |
20 μL | |
5976 | |
IgG1 |