Learn More
Unity Lab Services™ Premium Pipette Support Plan
Fast, quality-minded service that meets your diverse needs
84.55€ - 229.00€
Code produit | Marque | Service Offering | À utiliser avec (équipement) | Prix | Quantité et disponibilité | ||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Service Offering | À utiliser avec (équipement) | Prix | Quantité et disponibilité | ||||
16369386
|
Unity Lab Services™
GXP-3X10-DMC |
GxP Preventive Maintenance and Calibration, 3 x 10 | Multichannel Pipette (Any Brand and Model) |
229.00€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17168208
|
Unity Lab Services™
GXP3X10DSC |
GxP Preventive Maintenance and Calibration, 3 x 10 | Single-channel Pipette (Any Brand and Model) |
84.55€
1 pièce |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Whether cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette brands and models. We provide calibration and a detailed 15-point pipette inspection of single and multichannel manual and electronic components to identify potential concerns that can impact pipettes from performing optimally. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Unity Pipette Services offers:
- Service for any pipette brand and model
- Reliable service options by our highly trained technicians
- Fast and convenient prepaid pipette shipping packages
Premium – GxP Preventive Maintenance and Calibration
The GxP Preventive Maintenance and Calibration Plan provides a comprehensive service that includes gravimetric evaluation of your pipette's performance prior to any inspection, preventive maintenance or minor repairs. This plan includes a calibration certificate and sticker.
Ideal for customers who require:
- As-received data
- Pipette inspection, liquid leak test, routine preventive maintenance and minor repairs
- Calibration
- Work in a highly regulated environment
- Reduced OOTs (Out of Tolerance) that could invalidate research