Learn More
UBE2S Mouse anti-Human, Unconjugated, Clone: 8C9E6, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66897-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The UBE2S antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBE2S. This antibody recognizes human antigen. The UBE2S antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
UBE2S | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
8C9E6 | |
Unconjugated | |
20 μL | |
27338 | |
IgG2b |