Learn More
TSC22D2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25418-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TSC22D2 belongs to the TSC-22/Dip/Bun family and the function remains unknown.Spécification
TSC22D2 | |
Polyclonal | |
Unconjugated | |
TSC22D2 | |
KIAA0669, TILZ4, TILZ4a, TILZ4b, TILZ4c, TSC22 domain family protein 2, TSC22 domain family, member 2, TSC22D2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9819 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75157 | |
TSC22D2 | |
TSC22D2 Fusion Protein Ag22294 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |