Learn More
TPCN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23758-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TPCN1 may function as one of the major voltage-gated Ca2+ channel.Spécification
TPCN1 | |
Polyclonal | |
Unconjugated | |
TPCN1 | |
KIAA1169, TPC1, TPCN1, two pore segment channel 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
252972, 53373 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EQJ0, Q9ULQ1 | |
TPCN1 | |
TPCN1 Fusion Protein Ag20607 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |