missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TALDO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | TALDO1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16870064
|
Proteintech
12376-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16860064
|
Proteintech
12376-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.Spécification
TALDO1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P37837, Q93092, Q9EQS0 | |
21351, 6888, 83688 | |
TALDO1 Fusion Protein Ag3031 | |
Primary | |
-20°C | |
TALDO1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
TAL, TAL H, TALDO, TALDO1, TALDOR, TALH, Transaldolase, transaldolase 1 | |
Taldo1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |