missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TALDO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12376-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.Spécification
TALDO1 | |
Polyclonal | |
Unconjugated | |
TALDO1 | |
TAL, TAL H, TALDO, TALDO1, TALDOR, TALH, Transaldolase, transaldolase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21351, 6888, 83688 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P37837, Q93092, Q9EQS0 | |
Taldo1 | |
TALDO1 Fusion Protein Ag3031 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |