missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25265-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Plays a role in pre-mRNA alternative splicing by regulating spliceosome assembly.Specifica
SYNE2 | |
Polyclonal | |
Unconjugated | |
SYNE2 | |
KIAA1011, Nesprin 2, NUA, NUANCE, Protein NUANCE, SYNE 2, SYNE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23224 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8WXH0 | |
SYNE2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |