Learn More
Nitrate de sodium, 99,0 % min., qualité ACS, Alfa Aesar™
31.50€ - 217.00€
Products 4
Produktcode Marke Quantité Preis Menge & Verfügbarkeit  
Produktcode Marke Quantité Preis Menge & Verfügbarkeit  
11362448
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014493.14
31.50€
25g
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
11342448
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014493.30
250g
39.90€
250g
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
N/A
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
11352448
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014493.A1
1kg
69.10€
1kg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
11332448
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
014493.A7
217.00€
5kg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Beschreibung
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
RUO – Research Use Only