missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrosulfide Hydrate, Honeywell Fluka™
min. 60.0% (NaHS)
19.23€ - 124.18€
Identifiants chimiques
CAS | 207683-19-0 |
---|---|
Formule moléculaire | HNaS |
Poids moléculaire (g/mol) | 56.06 |
Numéro MDL | MFCD00149796 |
Clé InChI | HYHCSLBZRBJJCH-UHFFFAOYSA-M |
Synonyme | sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp |
CID PubChem | 16211346 |
SMILES | [Na+].[SH-] |
Code produit | Marque | Quantité | Conditionnement | Prix | Quantité et disponibilité | ||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Conditionnement | Prix | Quantité et disponibilité | ||||
15611540
|
Honeywell-Fluka
161527-5G |
5 g | Glass Bottle | ||||||
15681530
|
Honeywell-Fluka
161527-100G |
100 g | Glass Bottle | ||||||
15691530
|
Honeywell-Fluka
161527-1KG |
1 kg | Glass Bottle | ||||||
15601540
|
Honeywell-Fluka
161527-3KG |
3 kg | Glass Bottle | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Identifiants chimiques
207683-19-0 | |
56.06 | |
HYHCSLBZRBJJCH-UHFFFAOYSA-M | |
16211346 |
HNaS | |
MFCD00149796 | |
sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp | |
[Na+].[SH-] |
Spécification
Sodium Hydrosulfide Hydrate | |
207683-19-0 | |
HNaS | |
MFCD00149796 | |
90°C (194°F) | |
HYHCSLBZRBJJCH-UHFFFAOYSA-M | |
sodium;sulfanide;hydrate | |
16211346 | |
Flakes |
52°C to 54°C | |
Yellow | |
NaSH·xH2O | |
UN2949 | |
sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp | |
[Na+].[SH-] | |
56.06 | |
56.06g/mol (anhydrous basis) |
Sécurité et traitement
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314
missing translation for 'dotInformation' : 8/PGII