missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC20A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12423-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Retrovirus receptors allow infection of human and murine cells by various retroviruses. The receptors that have been identified at the molecular level include CD4 for human immunodeficiency virus, Rec1 for murine ecotropic virus, and GLVR1 for gibbon ape leukemia virus. These 3 proteins show no homology to one another at the DNA or protein level. GLVR1 is a sodium-dependent phosphate symporter.Spécification
SLC20A1 | |
Polyclonal | |
Unconjugated | |
SLC20A1 | |
Glvr 1, GLVR1, Phosphate transporter 1, PiT 1, PIT1, SLC20A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20515, 6574, 81826 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q61609, Q8WUM9, Q9JJP0 | |
SLC20A1 | |
SLC20A1 Fusion Protein Ag3084 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |