missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | RNF11 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16836744
|
Proteintech
16948-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16826744
|
Proteintech
16948-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Essential component of a ubiquitin-editing protein complex, comprising also TNFAIP3, ITCH and TAX1BP1, that ensures the transient nature of inflammatory signaling pathways. Promotes the association of TNFAIP3 to RIPK1 after TNF stimulation. TNFAIP3 deubiquitinates ′Lys-63′ polyubiquitin chains on RIPK1 and catalyzes the formation of ′Lys-48′-polyubiquitin chains. This leads to RIPK1 proteasomal degradation and consequently termination of the TNF- or LPS-mediated activation of NF-kappa-B. Recruits STAMBP to the E3 ubiquitin-ligase SMURF2 for ubiquitination, leading to its degradation by the 26S proteasome.Spécification
RNF11 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q9QYK7, Q9Y3C5 | |
26994, 29864, 689577 | |
RNF11 Fusion Protein Ag10563 | |
Primary | |
-20°C | |
RNF11 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CGI 123, ring finger protein 11, RNF11, SID1669 | |
Rnf11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |