Learn More
RLBP1L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21394-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3, 5-bisphosphate (PtdIns(3, 5)P2).Spécification
RLBP1L2 | |
Polyclonal | |
Unconjugated | |
CLVS2 | |
C6orf212, C6orf213, Clavesin 2, CLVS2, RLBP1L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134829, 215890, 361459 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A6JUQ6, Q5SYC1, Q8BG92 | |
Clvs2 | |
RLBP1L2 Fusion Protein Ag16061 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |