missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM42 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15407-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Binds (via the RRM domain) to the 3′-untranslated region (UTR) of CDKN1A mRNA.Spécification
RBM42 | |
Polyclonal | |
Unconjugated | |
RBM42 | |
RBM42, RNA binding motif protein 42, RNA binding protein 42 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361545, 68035, 79171 | |
-20°C | |
Liquid |
Western Blot | |
0.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AXT7, Q91V81, Q9BTD8 | |
Rbm42 | |
RBM42 Fusion Protein Ag7667 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |