missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB27B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13412-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RAB27B may be involved in targeting uroplakins to urothelial apical membranes.Spécification
RAB27B | |
Polyclonal | |
Unconjugated | |
RAB27B | |
C25KG, RAB27B, Ras related protein Rab 27B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5874, 80718, 84590 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00194, Q99P58, Q99P74 | |
Rab27b | |
RAB27B Fusion Protein Ag4064 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |