Learn More
PSPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14513-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome.Spécification
PSPH | |
Polyclonal | |
Unconjugated | |
PSPH | |
L 3 phosphoserine phosphatase, phosphoserine phosphatase, PSP, PSPase, PSPH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100678, 304429, 5723 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78330, Q5M819, Q99LS3 | |
PSPH | |
PSPH Fusion Protein Ag5972 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |