missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | PSPH |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16813184
|
Proteintech
14513-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16803184
|
Proteintech
14513-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome.Spécification
PSPH | |
Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P78330, Q5M819, Q99LS3 | |
100678, 304429, 5723 | |
PSPH Fusion Protein Ag5972 | |
Primary | |
-20°C | |
PSPH |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
L 3 phosphoserine phosphatase, phosphoserine phosphatase, PSP, PSPase, PSPH | |
PSPH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |