missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55143-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The antibody is specific to PSMB11.
Proteasomes generate peptides that are presented by major histocompatibility complex. Three additional subunits, beta-1i, beta-2i, and beta-5i, are induced by gamma-interferon and are preferentially incorporated into proteasomes to make immunoproteasomes. PSMB11, or beta-5t, is a catalytic subunit expressed exclusively in cortical thymic epithelial cells.Spécification
PSMB11 | |
Polyclonal | |
Unconjugated | |
PSMB11 | |
beta5t, Proteasome subunit beta 5t, PSMB11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122706, 73902 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A5LHX3, Q8BG41 | |
Psmb11 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |