missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15414-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.Spécification
PRPF19 | |
Polyclonal | |
Unconjugated | |
PRPF19 | |
hPSO4, Nuclear matrix protein 200, Pre mRNA processing factor 19, PRP19, PRP19/PSO4 homolog, PRPF19, PSO4, Senescence evasion factor, SNEV, UBOX4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246216, 27339, 28000 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99KP6, Q9JMJ4, Q9UMS4 | |
PRPF19 | |
PRPF19 Fusion Protein Ag7700 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |