missing translation for 'onlineSavingsMsg'
Learn More

Prohibitin Mouse anti-Human, Porcine, Rat, Clone: 4D3G5, Proteintech

Mouse Monoclonal Antibody

145.00€ - 399.00€

Spécification

Antigène Prohibitin
Clone 4D3G5
Concentration 0.8 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16837065
Afficher les documents
Proteintech
60092-1-IG-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16827065
Afficher les documents
Proteintech
60092-1-IG-150UL
150 μL
399.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Prohibitin is evolutionarily conserved, and its product is proposed to play a role in human cellular senescence and tumor suppression. Antiproliferative activity is reported to be localized to the 3′ UTR, which is proposed to function as a trans-acting regulatory RNA. Several pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants.
Spécification

Spécification

Prohibitin
0.8 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PHB, PHB1, prohibitin
Phb
IgG1
Caprylic Acid Precipitation
Antibody
4D3G5
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Mouse
Porcine, Rat, Human
P35232, P67779
100524707, 25344, 5245
Prohibitin Fusion Protein Ag1240
Primary
-20°C
PHB
Vidéos
FDS
Documents

Documents

Product Certifications