Learn More
Prohibitin Mouse anti-Human, Porcine, Rat, Clone: 4D3G5, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60092-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Prohibitin is evolutionarily conserved, and its product is proposed to play a role in human cellular senescence and tumor suppression. Antiproliferative activity is reported to be localized to the 3′ UTR, which is proposed to function as a trans-acting regulatory RNA. Several pseudogenes of this gene have been identified. Alternative splicing results in multiple transcript variants.Spécification
Prohibitin | |
Monoclonal | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P35232, P67779 | |
Phb | |
Prohibitin Fusion Protein Ag1240 | |
20 μL | |
Primary | |
Porcine, Rat, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
4D3G5 | |
Unconjugated | |
PHB | |
PHB, PHB1, prohibitin | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
100524707, 25344, 5245 | |
-20°C | |
Liquid |