missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25114-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.Spécification
POU3F4 | |
Polyclonal | |
Unconjugated | |
POU3F4 | |
BRAIN 4, Brn 4, BRN4, DFN3, DFNX2, Oct 9, Octamer binding protein 9, OTF 9, OTF9, POU class 3 homeobox 4, POU3F4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18994, 5456 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49335, P62515 | |
Pou3f4 | |
POU3F4 Fusion Protein Ag18925 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |