Learn More
POU3F4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 25114-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16833655
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.
Spécification
Spécification
POU3F4
Polyclonal
Unconjugated
POU3F4
BRAIN 4, Brn 4, BRN4, DFN3, DFNX2, Oct 9, Octamer binding protein 9, OTF 9, OTF9, POU class 3 homeobox 4, POU3F4
Rabbit
Antigen Affinity Chromatography
RUO
18994, 5456
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49335, P62515
Pou3f4
POU3F4 Fusion Protein Ag18925
150 μL
Primary
Human, Mouse