Learn More
PHYHIPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16937-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
16937-1-AP detects 39 kDa and 42-45 kDa isoforms in SDS-PAGE.
PHYHIPL may play a role in the development of the central system.Spécification
PHYHIPL | |
Polyclonal | |
Unconjugated | |
PHYHIPL | |
KIAA1796, PHYHIPL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309901, 70911, 84457 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AYN4, Q8BGT8, Q96FC7 | |
Phyhipl | |
PHYHIPL Fusion Protein Ag10435 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |