missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55197-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Protects DRG2 from proteolytic degradation.Spécification
PAPD5 | |
Polyclonal | |
Unconjugated | |
PAPD5 | |
PAPD5, Terminal uridylyltransferase 3, TRF4 2, TUTase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214627, 64282 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68ED3, Q8NDF8 | |
PAPD5 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |