Learn More
P2RX7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène P2RX7
Concentration 300 μg/mL
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications
Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16806055
Afficher les documents
Proteintech
28207-1-AP-20UL
20 μL
145.00€
20µL
16896045
Afficher les documents
Proteintech
28207-1-AP-150UL
150 μL
410.00€
150µL
Description
Description
The P2RX7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human P2RX7. This antibody recognizes human, mouse antigen. The P2RX7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
P2RX7
Western Blot, Immunohistochemistry
Unconjugated
Human, Mouse
IgG
300 μg/mL
Polyclonal
Rabbit
5027