Learn More
P2RX7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28207-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The P2RX7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human P2RX7. This antibody recognizes human, mouse antigen. The P2RX7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
P2RX7 | |
Polyclonal | |
Unconjugated | |
150 μL | |
5027 | |
IgG |
Western Blot, Immunohistochemistry | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |