missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5M Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20765-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Dephosphorylates specifically the 5′ and 2′(3′)- phosphates of uracil and thymine deoxyribonucleotides, and so protects mitochondrial DNA replication from excess dTTP. Has only marginal activity towards dIMP and dGMP.Spécification
NT5M | |
Polyclonal | |
Unconjugated | |
NT5M | |
Deoxy 5 nucleotidase 2, dNT 2, dNT2, mdN, NT5M | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103850, 287368, 56953 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VCE6, Q9NPB1 | |
Nt5m | |
NT5M Fusion Protein Ag14743 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |